Key Documents
B9182
Sigma-Aldrich
α-D-6-O-Phosphomannopyranoside polyacrylamide biotin
Synonym(s):
α-D-Man-6-P-PAA-biotin
Sign Into View Organizational & Contract Pricing
About This Item
MDL number:
UNSPSC Code:
12352201
mol wt
~30 kDa
Choose from one of the most recent versions:
Certificates of Analysis (COA)